1-Isopropylpiperazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

566853-1G
SIAL566853-1GEA 27 EUR
SIAL566853-1G
1-Isopropylpiperazin, Sigma-Aldrich®
1-Isopropylpiperazin
Formel: C₇H₁₆N₂
Molecular Weight: 128,22 g/mol
MDL: MFCD00167971
CAS-Nummer: 4318-42-7
EINECS: 224-345-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR