4-Brombenzylmercaptan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: p-Bromtoluol-α-thiol

566950-1G
SIAL566950-1GEA 64.3 EUR
SIAL566950-1G
4-Brombenzylmercaptan, Sigma-Aldrich®
4-Brombenzylmercaptan
Formel: C₇H₇BrS
Molecular Weight: 203,1 g/mol
MDL: MFCD03701566
CAS-Nummer: 19552-10-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR