Tetramethylthiurammonosulfid
Lieferant: Merck
|
Achtung
|
Synonyme:
Bis(dimethylthiocarbamyl) sulphide
M-Clarity™ Qualitätsstufe = MQ100
567205-100G
SIAL567205-100GEA
92.4
EUR
SIAL567205-100G
Tetramethylthiurammonosulfid
Tetramethylthiurammonosulfid
Formel:
C₆H₁₂N₂S₃ Molecular Weight: 208,37 g/mol Schmelzpunkt: 110 °C Dichte: 1,37 g/cm³ (20 °C) |
MDL:
MFCD00014870 CAS-Nummer: 97-74-5 EINECS: 202-605-7 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...