N-(3-Hydroxypropyl)morpholin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Morpholinopropanol, 3-Morpholino-1-propanol, 4-(3-Hydroxypropyl)morpholine, 3-(4-Morpholinyl)-1-propanol

M-Clarity™ Qualitätsstufe = MQ100

567280-1G
SIAL567280-1GEA 57.8 EUR
SIAL567280-1G
N-(3-Hydroxypropyl)morpholin, Sigma-Aldrich®
N-(3-Hydroxypropyl)morpholin
Formel: C₇H₁₅NO₂
Molecular Weight: 145,2 g/mol
MDL: MFCD01713533
CAS-Nummer: 4441-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR