5-Fluor-2-methylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-5-fluorophenylboronic acid

M-Clarity™ Qualitätsstufe = MQ100

567442-1G
SIAL567442-1GEA 52.3 EUR
SIAL567442-1G
5-Fluor-2-methylphenylboronsäure, Sigma-Aldrich®
5-Fluor-2-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,95 g/mol
MDL: MFCD03095047
CAS-Nummer: 163517-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR