2-Chlor-4-(trifluormethyl)benzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlor-4-(trifluormethyl)benzoesäurenitril, 3-Chlor-4-cyanbenzotrifluorid

M-Clarity™ Qualitätsstufe = MQ200

567620-1G
SIAL567620-1GEA 51.4 EUR
SIAL567620-1G
2-Chlor-4-(trifluormethyl)benzonitril, Sigma-Aldrich®
2-Chlor-4-(trifluormethyl)benzonitril
Formel: C₈H₃ClF₃N
Molecular Weight: 205,57 g/mol
MDL: MFCD00084939
CAS-Nummer: 1813-33-8
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR