3-(2,5-Dimethoxybenzoyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

568244-5G
SIAL568244-5GEA 112 EUR
SIAL568244-5G
3-(2,5-Dimethoxybenzoyl)propionsäure, Sigma-Aldrich®
3-(2,5-Dimethoxybenzoyl)propionsäure
Formel: C₁₂H₁₄O₅
MDL: MFCD00089268
CAS-Nummer: 1084-74-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR