Ethyl-3-chlor-4-hydroxybenzoat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

568325-5G
SIAL568325-5GEA 254 EUR
SIAL568325-5G
Ethyl-3-chlor-4-hydroxybenzoat, Sigma-Aldrich®
Ethyl-3-chlor-4-hydroxybenzoat
Formel: C₉H₉ClO₃
Molecular Weight: 200,62 g/mol
MDL: MFCD05664241
CAS-Nummer: 16357-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR