trans-3-Methoxy-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

568686-5G
SIAL568686-5GEA 80.7 EUR
SIAL568686-5G
trans-3-Methoxy-β-nitrostyrol, Sigma-Aldrich®
trans-3-Methoxy-β-nitrostyrol
Formel: C₉H₉NO₃
Molecular Weight: 179,18 g/mol
MDL: MFCD00024822
CAS-Nummer: 3179-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR