16-Brompalmitinsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

568708-5G
SIAL568708-5GEA 493 EUR
SIAL568708-5G
16-Brompalmitinsäure, Sigma-Aldrich®
16-Brompalmitinsäure
Formel: BrH₂C(CH₂)₁₄COOH
Lagertemperatur: Raumtemperatur
MDL: MFCD00040779
CAS-Nummer: 2536-35-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR