1,3-Phenylendiisothiocyanat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

568937-1G
SIAL568937-1GEA 237 EUR
SIAL568937-1G
1,3-Phenylendiisothiocyanat, Sigma-Aldrich®
1,3-Phenylendiisothiocyanat
Formel: C₈H₄N₂S₂
Molecular Weight: 192,27 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 3125-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR