4-Chlor-2-fluorphenylisocyanat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

569100-1G
SIAL569100-1GEA 123 EUR
SIAL569100-1G
4-Chlor-2-fluorphenylisocyanat, Sigma-Aldrich®
4-Chlor-2-fluorphenylisocyanat
Formel: C₇H₃ClFNO
Molecular Weight: 171,56 g/mol
CAS-Nummer: 51163-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR