4,6-Dichlor-o-toluidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Dichloro-6-methylaniline

M-Clarity™ Qualitätsstufe = MQ100

569240-5G
SIAL569240-5GEA 61.1 EUR
SIAL569240-5G
4,6-Dichlor-o-toluidin, Sigma-Aldrich®
4,6-Dichlor-o-toluidin
Formel: C₇H₇Cl₂N
Molecular Weight: 176,05 g/mol
MDL: MFCD00044074
CAS-Nummer: 30273-00-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR