3-[(Trimethylsilyl)ethinyl]anisol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

569321-5G
SIAL569321-5GEA 102 EUR
SIAL569321-5G
3-[(Trimethylsilyl)ethinyl]anisol, Sigma-Aldrich®
3-[(Trimethylsilyl)ethinyl]anisol
Formel: C₁₂H₁₆OSi
MDL: MFCD03701606
CAS-Nummer: 40230-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR