Indigocarmin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Acid Blue 74, Food blue No. 2
57000-100G-F
57000-25G-F
57000-500G-F
SIAL57000-100G-FEA
141
EUR
SIAL57000-100G-F
SIAL57000-25G-F
SIAL57000-500G-F
Indigocarmin, Sigma-Aldrich®
Indigocarmin
Formel:
C₁₆H₈N₂Na₂O₈S₂ Schmelzpunkt: > 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005723 CAS-Nummer: 860-22-0 EINECS: 212-728-8 Merck Index: 13,04969 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...