1-(2-Tetrahydrofuroyl)piperazin Hydrobromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Piperazin-1-yl(tetrahydrofuran-2-yl)methanone hydrobromide

M-Clarity™ Qualitätsstufe = MQ100

570990-5G
SIAL570990-5GEA 122 EUR
SIAL570990-5G
1-(2-Tetrahydrofuroyl)piperazin Hydrobromid, Sigma-Aldrich®
1-(2-Tetrahydrofuroyl)piperazin Hydrobromid
Formel: C₉H₁₇BrN₂O₂
Molecular Weight: 265,1475 g/mol
MDL: MFCD03701619
CAS-Nummer: 63590-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR