Kalium(4-methylphenyl)trifluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Kalium(p-tolyl)trifluoroborat, (4-Methylphenyl)trifluorborsäure Kaliumsalz

M-Clarity™ Qualitätsstufe = MQ100

571555-1G
SIAL571555-1GEA 57.8 EUR
SIAL571555-1G
Kalium(4-methylphenyl)trifluoroborat, Sigma-Aldrich®
Kalium(4-methylphenyl)trifluoroborat
Formel: C₇H₇BF₃K
Molecular Weight: 198,04 g/mol
MDL: MFCD01318171
CAS-Nummer: 216434-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR