Tris(triphenylphosphin)kupfer(I)bromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bromotris(triphenylphosphine)copper(I)

M-Clarity™ Qualitätsstufe = MQ100

572144-5G
SIAL572144-5GEA 60.3 EUR
SIAL572144-5G
Tris(triphenylphosphin)kupfer(I)bromid, Sigma-Aldrich®
Tris(triphenylphosphin)kupfer(I)bromid
Formel: C₅₄H₄₅BrCuP₃
Molecular Weight: 930,32 g/mol
MDL: MFCD04118115
CAS-Nummer: 15709-74-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR