4-(2-Pyridyl)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Pyridin-2-yl-benzaldehyde, 4-(Pyridin-2-yl)benzaldehyde

572411-5G
SIAL572411-5GEA 33 EUR
SIAL572411-5G
4-(2-Pyridyl)benzaldehyd, Sigma-Aldrich®
4-(2-Pyridyl)benzaldehyd
Formel: C₁₂H₉NO
Molecular Weight: 183,21 g/mol
MDL: MFCD01863537
CAS-Nummer: 127406-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR