Germanium(II)bromid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

572659-5G
SIAL572659-5GEA 247 EUR
SIAL572659-5G
Germanium(II)bromid, Sigma-Aldrich®
Germanium(II) bromide
Formel: Br₂Ge
Molecular Weight: 232,42 g/mol
CAS-Nummer: 24415-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR