3-Amino-3-(4-chlorphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Amino-3-(4-chlorophenyl)propanoic acid, 3-Amino-3-(4-chlorophenyl)propionic Acid, 3-Amino-3-(p-chlorophenyl)propionic acid

M-Clarity™ Qualitätsstufe = MQ100

573353-1G
SIAL573353-1GEA 44.8 EUR
SIAL573353-1G
3-Amino-3-(4-chlorphenyl)propionsäure, Sigma-Aldrich®
3-Amino-3-(4-chlorphenyl)propionsäure
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,64 g/mol
MDL: MFCD00126370
CAS-Nummer: 19947-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR