3-Amino-3-(4-nitrophenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

573396-1G
SIAL573396-1GEA 48.6 EUR
SIAL573396-1G
3-Amino-3-(4-nitrophenyl)propionsäure, Sigma-Aldrich®
3-Amino-3-(4-nitrophenyl)propionsäure
Formel: C₉H₁₀N₂O₄
MDL: MFCD00187210
CAS-Nummer: 35005-61-9
EINECS: 252-316-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR