3-Amino-3-(4-fluorphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

573418-1G
SIAL573418-1GEA 43.5 EUR
SIAL573418-1G
3-Amino-3-(4-fluorphenyl)propionsäure, Sigma-Aldrich®
3-Amino-3-(4-fluorphenyl)propionsäure
Formel: C₉H₁₀FNO₂
Molecular Weight: 183,18 g/mol
MDL: MFCD00181811
CAS-Nummer: 325-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR