Cer(III)sulfat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dicertrisulfat

M-Clarity™ Qualitätsstufe = MQ100

574201-25G
SIAL574201-25GEA 215 EUR
SIAL574201-25G
Cer(III)sulfat, Sigma-Aldrich®
Cerium(III) sulphate
Formel: Ce₂O₁₂S₃
Molecular Weight: 568,42 g/mol
Dichte: 2,89 g/cm³ (20 °C)
CAS-Nummer: 13454-94-9
EINECS: 236-644-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR