3,5-Dichlor-2-pyridon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Dichloro-2-hydroxypyridine

M-Clarity™ Qualitätsstufe = MQ100

574414-1G
SIAL574414-1GEA 71.8 EUR
SIAL574414-1G
3,5-Dichlor-2-pyridon, Sigma-Aldrich®
3,5-Dichlor-2-pyridon
Formel: C₅H₃Cl₂NO
Molecular Weight: 163,99 g/mol
MDL: MFCD00015401
CAS-Nummer: 5437-33-2
EINECS: 226-609-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR