5-Bromnicotinonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Brom-5-pyridincarbonitril, 5-Bromnicotinsäurenitril, 3-Brom-5-pyridincarbonsäurenitril, 3-Brom-5-cyanpyridin

M-Clarity™ Qualitätsstufe = MQ100

574422-1G
SIAL574422-1GEA 45.4 EUR
SIAL574422-1G
5-Bromnicotinonitril, Sigma-Aldrich®
5-Bromnicotinonitril
Formel: C₆H₃BrN₂
Molecular Weight: 183,01 g/mol
MDL: MFCD00174363
CAS-Nummer: 35590-37-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR