1-Butyl-3-methylimidazoliumhydrogensulfat, Sigma-Aldrich®

Lieferant: Merck

57457-1KG-F 57457-100G-F
SIAL57457-1KG-FEA 1930 EUR
SIAL57457-1KG-F SIAL57457-100G-F
1-Butyl-3-methylimidazoliumhydrogensulfat, Sigma-Aldrich®
1-Butyl-3-methylimidazolium hydrogen sulphate
Formel: C₈H₁₆N₂O₄S
MDL: MFCD06798197
CAS-Nummer: 262297-13-2
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR