2,5-Dibromhydrochinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Dibromobenzene-1,4-diol

574619-25G 574619-5G
SIAL574619-25GEA 245 EUR
SIAL574619-25G SIAL574619-5G
2,5-Dibromhydrochinon, Sigma-Aldrich®
2,5-Dibromhydrochinon
Formel: C₆H₄Br₂O₂
Molecular Weight: 267.9 g/mol
MDL: MFCD00192664
CAS-Nummer: 14753-51-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR