4-(Ethoxycarbonyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid

M-Clarity™ Qualitätsstufe = MQ100

574643-1G 574643-5G
SIAL574643-1GEA 68.3 EUR
SIAL574643-1G SIAL574643-5G
4-(Ethoxycarbonyl)phenylboronsäure, Sigma-Aldrich®
4-(Ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₁BO₄
MDL: MFCD02179441
CAS-Nummer: 4334-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR