3-Carboxyphenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Carboxybenzeneboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

M-Clarity™ Qualitätsstufe = MQ100

574694-1G
SIAL574694-1GEA 31.7 EUR
SIAL574694-1G
3-Carboxyphenylboronsäurepinakolester, Sigma-Aldrich®
3-Carboxyphenylboronsäurepinakolester
Formel: C₁₃H₁₇BO₄
Molecular Weight: 248,083 g/mol
MDL: MFCD03411930
CAS-Nummer: 269409-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR