3,5-Dichlorisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Dichloropyridine-4-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

574880-5G
SIAL574880-5GEA 76.3 EUR
SIAL574880-5G
3,5-Dichlorisonicotinsäure, Sigma-Aldrich®
3,5-Dichlorisonicotinsäure
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192 g/mol
MDL: MFCD01861975
CAS-Nummer: 13958-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR