Ammoniumnickel(II)sulfat Hexahydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ammonium nickel sulphate hexahydrate, Nickel ammonium sulphate Ammonium disulphatonickelate, Nickel ammonium sulphate hexahydrate

M-Clarity™ Qualitätsstufe = MQ100

574988-25G
SIAL574988-25GEA 196 EUR
SIAL574988-25G
Ammoniumnickel(II)sulfat Hexahydrat, Sigma-Aldrich®
Ammoniumnickel(II)sulfat Hexahydrat
Formel: H₈N₂NiO₈S₂·6H₂O
Molecular Weight: 394,99 g/mol
MDL: MFCD00150260
CAS-Nummer: 7785-20-8
EINECS: 239-793-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR