Lanthan(III)sulfat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dilanthantrisulfat

M-Clarity™ Qualitätsstufe = MQ100

575208-50G
SIAL575208-50GEA 208 EUR
SIAL575208-50G
Lanthan(III)sulfat, Sigma-Aldrich®
Lanthan(III)sulfat
Formel: La₂O₁₂S₃
Molecular Weight: 566 g/mol
MDL: MFCD00011070
CAS-Nummer: 10099-60-2
EINECS: 233-239-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR