Europium(III)bromid Hydrat, Sigma-Aldrich®

Lieferant: Merck

575259-5G
SIAL575259-5GEA 83.7 EUR
SIAL575259-5G
Europium(III)bromid Hydrat, Sigma-Aldrich®
Europium(III)bromid Hydrat
Formel: Br₃Eu·H₂O
Molecular Weight: 409,69 g/mol
MDL: MFCD00049921
CAS-Nummer: 560069-78-5
EINECS: 237-349-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR