3-(2,4-Dimethoxyphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(2,4-Dimethoxyphenyl)propanoic acid

M-Clarity™ Qualitätsstufe = MQ100

575666-5G
SIAL575666-5GEA 150 EUR
SIAL575666-5G
3-(2,4-Dimethoxyphenyl)propionsäure, Sigma-Aldrich®
3-(2,4-Dimethoxyphenyl)propionsäure
Formel: C₁₁H₁₄O₄
Molecular Weight: 210,23 g/mol
MDL: MFCD00598205
CAS-Nummer: 22174-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR