1-Methylimidazol-2-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Methyl-1H-imidazole-2-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

576220-1G
SIAL576220-1GEA 43.3 EUR
SIAL576220-1G
1-Methylimidazol-2-carbonsäure, Sigma-Aldrich®
1-Methylimidazol-2-carbonsäure
Formel: C₅H₆N₂O₂
Molecular Weight: 126,11 g/mol
MDL: MFCD01863421
CAS-Nummer: 20485-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR