Lithiumbis(trimethylsilyl)amid 1 mol/l in tert-Butylmethylether, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Lithiumbis(trimethylsilyl)amid, HMDS Lithiumsalz, N-Lithiohexamethyldisilazan, HMDS-Li
M-Clarity™ Qualitätsstufe = MQ100
577014-100ML
SIAL577014-100MLEA
145
EUR
SIAL577014-100ML
Lithiumbis(trimethylsilyl)amid 1 mol/l in tert-Butylmethylether, Sigma-Aldrich®
Lithiumbis(trimethylsilyl)amid
Formel:
C₆H₁₈LiNSi₂ Molecular Weight: 167,33 g/mol |
MDL:
MFCD00008261 CAS-Nummer: 4039-32-1 EINECS: 223-725-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...