1,1'-Bis(diisopropylphosphino)ferrocen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: DiPPF, 1,1'-Bis(di-i-propylphosphino)ferrocene

M-Clarity™ Qualitätsstufe = MQ100

578266-500MG
SIAL578266-500MGEA 164 EUR
SIAL578266-500MG
1,1'-Bis(diisopropylphosphino)ferrocen, Sigma-Aldrich®
1,1'-Bis(diisopropylphosphino)ferrocen
Formel: C₂₂H₂₈FeP₂
Molecular Weight: 410.25 g/mol
MDL: MFCD02684559
CAS-Nummer: 97239-80-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR