Methyl-6-indolcarboxylat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Methyl 1H-indole-6-carboxylate
M-Clarity™ Qualitätsstufe = MQ100
578495-5G
SIAL578495-5GEA
57.3
EUR
SIAL578495-5G
Methyl-6-indolcarboxylat, Sigma-Aldrich®
Methyl-6-indolcarboxylat
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol |
MDL:
MFCD00211063 CAS-Nummer: 50820-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...