3-Ethinylthiophen, Sigma-Aldrich®

Lieferant: Merck

578797-1G 578797-5G
SIAL578797-1GEA 76.8 EUR
SIAL578797-1G SIAL578797-5G
3-Ethinylthiophen, Sigma-Aldrich®
3-Ethynylthiophene
Formel: C₆H₄S
Molecular Weight: 108.16 g/mol
MDL: MFCD04039973
CAS-Nummer: 67237-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR