3-(Boc-amino)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: tert-Butyl (3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate, 3-(Boc-amino)benzeneboronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

578843-1G
SIAL578843-1GEA 78.1 EUR
SIAL578843-1G
3-(Boc-amino)phenylboronsäurepinakolester, Sigma-Aldrich®
3-(Boc-amino)phenylboronsäurepinakolester
Formel: C₁₇H₂₆BNO₄
Molecular Weight: 319,21 g/mol
MDL: MFCD03789256
CAS-Nummer: 330793-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR