Tri-tert-butylphosphoniumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tri-t-butylphosphonium tetrafluoroborate

M-Clarity™ Qualitätsstufe = MQ100

578940-1G 578940-5G
SIAL578940-1GEA 98.1 EUR
SIAL578940-1G SIAL578940-5G
Tri-tert-butylphosphoniumtetrafluoroborat, Sigma-Aldrich®
Tri-tert-butylphosphoniumtetrafluoroborat
Formel: C₁₂H₂₈BF₄P
Molecular Weight: 290.13 g/mol
MDL: MFCD04039975
CAS-Nummer: 131274-22-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR