Kalium(4-methoxyphenyl)trifluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Methoxyphenyl)trifluorborsäure Kaliumsalz

579068-1G
SIAL579068-1GEA 65.3 EUR
SIAL579068-1G
Kalium(4-methoxyphenyl)trifluoroborat, Sigma-Aldrich®
Kalium(4-methoxyphenyl)trifluoroborat
Formel: C₇H₇BF₃KO
Molecular Weight: 214,04 g/mol
MDL: MFCD04112717
CAS-Nummer: 192863-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR