4-Methoxy-2-nitrobenzonitril, Sigma-Aldrich®

Lieferant: Merck

579130-5G
SIAL579130-5GEA 48.3 EUR
SIAL579130-5G
4-Methoxy-2-nitrobenzonitril, Sigma-Aldrich®
4-Methoxy-2-nitrobenzonitril
Formel: C₈H₆N₂O₃
Molecular Weight: 178,1500 g/mol
MDL: MFCD00667741
CAS-Nummer: 38469-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR