5-Amino-2-(trifluormethyl)benzimidazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

579483-1G
SIAL579483-1GEA 55 EUR
SIAL579483-1G
5-Amino-2-(trifluormethyl)benzimidazol, Sigma-Aldrich®
5-Amino-2-(trifluoromethyl)benzimidazole
Formel: C₈H₆F₃N₃
Molecular Weight: 201,15 g/mol
MDL: MFCD01632182
CAS-Nummer: 3671-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR