2,3-Difluorbenzamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3-Difluorbenzoesäureamid

M-Clarity™ Qualitätsstufe = MQ100

579491-5G
SIAL579491-5GEA 54.4 EUR
SIAL579491-5G
2,3-Difluorbenzamid, Sigma-Aldrich®
2,3-Difluorbenzamid
Formel: C₇H₅F₂NO
Molecular Weight: 157,12 g/mol
MDL: MFCD00061137
CAS-Nummer: 18355-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR