5-Amino-α,α,α-trifluor-m-toluylsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

579513-1G
SIAL579513-1GEA 63.5 EUR
SIAL579513-1G
5-Amino-α,α,α-trifluor-m-toluylsäure, Sigma-Aldrich®
5-Amino-α,α,α-trifluor-m-toluylsäure
Formel: C₈H₆F₃NO₂
Molecular Weight: 205,14 g/mol
MDL: MFCD00236641
CAS-Nummer: 328-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR