2-Acetyl-5-iodthiophen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

579637-1G
SIAL579637-1GEA 30.8 EUR
SIAL579637-1G
2-Acetyl-5-iodthiophen, Sigma-Aldrich®
2-Acetyl-5-iodthiophen
Formel: C₆H₅IOS
MDL: MFCD02093967
CAS-Nummer: 30955-94-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR