3-Cyancumarin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Coumarin-3-carbonitrile, 2-Oxo-2H-chromene-3-carbonitrile

M-Clarity™ Qualitätsstufe = MQ200

579696-1G
SIAL579696-1GEA 38.6 EUR
SIAL579696-1G
3-Cyancumarin, Sigma-Aldrich®
3-Cyancumarin
Formel: C₁₀H₅NO₂
Molecular Weight: 171,16 g/mol
MDL: MFCD00115699
CAS-Nummer: 15119-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR