4-Brom-3,5-dimethoxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

579718-25G
SIAL579718-25GEA 128 EUR
SIAL579718-25G
4-Brom-3,5-dimethoxybenzoesäure, Sigma-Aldrich®
4-Bromo-3,5-dimethoxybenzoic acid
Formel: C₉H₉BrO₄
Molecular Weight: 261.07 g/mol
MDL: MFCD01632140
CAS-Nummer: 56518-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR